Anti-NBEA Goat Polyclonal Antibody

Supplier: Everest Biotechnology
EVBIEB06147EA 317 GBP
EVBIEB06147
Anti-NBEA Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NBEA (neurobeachin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR