Anti-IGF1R Rabbit Polyclonal Antibody
Supplier: Novus Biologicals
Certificates
About this item
Rabbit Polyclonal IGF1 Receptor Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Primate.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: IGF1R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- NOVUNBP1-77680SS
- NOVUNBP1-77680
- Antigen symbol:
- IGF1R
- IGF1R
- Antigen name:
- Insulin-Like Growth Factor 1 Receptor
- Insulin-Like Growth Factor 1 Receptor
- Antigen synonyms:
- MGC18216|JTK13|CD221|IGFIR|MGC142170|IGF1R|MGC142172
- MGC18216|JTK13|CD221|IGFIR|MGC142170|IGF1R|MGC142172
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 3480
- 3480
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifications
- SizePk
- : NBP1-77680SS: NOVUNBP1-77680SS
Specifications
Antigen symbolIGF1RAntigen nameInsulin-Like Growth Factor 1 ReceptorEnvironmentally PreferableAntigen synonymsMGC18216|JTK13|CD221|IGFIR|MGC142170|IGF1R|MGC142172Antibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene ID3480IsotypeWestern blotYesImmunoFluorescenceYesStorage temperatureCoolShipping temperatureIce - : NBP1-77680: NOVUNBP1-77680
Specifications
Antigen symbolIGF1RAntigen nameInsulin-Like Growth Factor 1 ReceptorEnvironmentally PreferableAntigen synonymsMGC18216|JTK13|CD221|IGFIR|MGC142170|IGF1R|MGC142172Antibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHumanHostRabbitGene ID3480IsotypeWestern blotYesImmunoFluorescenceYesStorage temperatureCoolShipping temperatureIce