Anti-mtTFA Mouse Monoclonal Antibody [clone: 18G102B2E11]
Mouse Monoclonal mtTFA Antibody (18G102B2E11). Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:200
Type: Primary
Antigen: MTTF1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- NOVUNBP1-71648
- NOVUNBP1-71648SS
- Antigen symbol:
- TFAM
- TFAM
- Antigen name:
- Transcription Factor A, Mitochondrial
- Transcription Factor A, Mitochondrial
- Antigen synonyms:
- mitochondrial transcription factor A|mtTFA|TCF-6|TCF6L1|mitochondrial transcription factor 1|Transcription factor 6-like 2 (mitochondrial transcription factor)|TCF6L2|transcription factor 6|TCF6L3|TCF6|MtTF1
- mitochondrial transcription factor A|mtTFA|TCF-6|TCF6L1|mitochondrial transcription factor 1|Transcription factor 6-like 2 (mitochondrial transcription factor)|TCF6L2|transcription factor 6|TCF6L3|TCF6|MtTF1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 18G102B2E11
- 18G102B2E11
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 7019
- 7019
- Isotype:
- IgG2b kappa
- IgG2b kappa
- Western blot:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
Specifications
Antigen symbolTFAMAntigen nameTranscription Factor A, MitochondrialEnvironmentally PreferableAntigen synonymsmitochondrial transcription factor A|mtTFA|TCF-6|TCF6L1|mitochondrial transcription factor 1|Transcription factor 6-like 2 (mitochondrial transcription factor)|TCF6L2|transcription factor 6|TCF6L3|TCF6|MtTF1Antibody typePrimaryClonalityMonoclonalConjugationUnconjugatedClone18G102B2E11ReactivityHumanHostMouseGene ID7019IsotypeIgG2b kappaWestern blotYesImmunoFluorescenceYesStorage temperatureCoolShipping temperatureIceSpecifications
Antigen symbolTFAMAntigen nameTranscription Factor A, MitochondrialEnvironmentally PreferableAntigen synonymsmitochondrial transcription factor A|mtTFA|TCF-6|TCF6L1|mitochondrial transcription factor 1|Transcription factor 6-like 2 (mitochondrial transcription factor)|TCF6L2|transcription factor 6|TCF6L3|TCF6|MtTF1Antibody typePrimaryClonalityMonoclonalConjugationUnconjugatedClone18G102B2E11ReactivityHumanHostMouseGene ID7019IsotypeIgG2b kappaWestern blotYesImmunoFluorescenceYesStorage temperatureCoolShipping temperatureIce