Northern Ireland
Order Entry
Anti-AR Mouse monoclonal antibody unconjugated [clone: 2H8]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-AR Mouse monoclonal antibody unconjugated [clone: 2H8]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-47471
  • NOVUNBP1-47471SS
  • Antigen symbol:
  • AR
  • AR
  • Antigen name:
  • Androgen Receptor
  • androgen receptor
  • Antigen synonyms:
  • AR|Nuclear receptor subfamily 3 group C member 4|NR3C4|Dihydrotestosterone receptor|DHTR
  • AIS|AR8|TFM|HYSP1|SBMA|SMAX1|NR3C4|HUMARA|KD|DHTR
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2H8
  • 2H8
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 367
  • 367
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

 

  •  
     
    Size
    Pk
     
     
  • NOVUNBP1-47471
    NBP1-47471
    100 µL
    0,1 mL
       
       
    1 * 0,1 mL
    1 * 0,1 mL - £685.00
    £685.00
    1 * 0,1 mL
       £685.00

     : NBP1-47471
     : NOVUNBP1-47471
     
     
    Specifications
    Antigen symbol
    AR
    Antigen name
    Androgen Receptor
    Environmentally Preferable
    Antigen synonyms
    AR|Nuclear receptor subfamily 3 group C member 4|NR3C4|Dihydrotestosterone receptor|DHTR
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    2H8
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    367
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • NOVUNBP1-47471SS
    NBP1-47471SS
    25 μL
    25 µl
       
       
    1 * 25 µl
    1 * 25 µl - £212.00
    £212.00
    1 * 25 µl
       £212.00

     : NBP1-47471SS
     : NOVUNBP1-47471SS
     
     
    Specifications
    Antigen symbol
    AR
    Antigen name
    androgen receptor
    Environmentally Preferable
    Antigen synonyms
    AIS|AR8|TFM|HYSP1|SBMA|SMAX1|NR3C4|HUMARA|KD|DHTR
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    2H8
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    367
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice