Northern Ireland
Order Entry
Anti-TUBB3 Mouse Monoclonal Antibody [clone: 2E9]
Anti-TUBB3 Mouse Monoclonal Antibody [clone: 2E9]
 
undefined
Anti-TUBB3 Mouse Monoclonal Antibody [clone: 2E9]

Specifications

  • Catalog No:
  • NOVUNBP1-42568
  • NOVUNBP1-42568SS
  • Antigen symbol:
  • TUBB3
  • TUBB3
  • Antigen name:
  • Tubulin Beta 3
  • tubulin, beta 3 class III
  • Antigen synonyms:
  • CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
  • CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 2E9
  • 2E9
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 10381
  • 10381
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • NOVUNBP1-42568
    NBP1-42568
    100 µL
    0,1 mL
       
    Log in for availability
    1 * 0,1 mL
    1 * 0,1 mL - £564.00
    £564.00
    1 * 0,1 mL
       £564.00

    Supplier Article Number: NBP1-42568
    Local Article Number: NOVUNBP1-42568
    Specifications:
    More
    Specifications
    Antigen symbol
    TUBB3
    Antigen name
    Tubulin Beta 3
    Environmentally Preferable
    Antigen synonyms
    CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    2E9
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    10381
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • NOVUNBP1-42568SS
    NBP1-42568SS
    25 μL
    25 µl
       
    Log in for availability
    1 * 25 µl
    1 * 25 µl - £248.00
    £248.00
    1 * 25 µl
       £248.00

    Supplier Article Number: NBP1-42568SS
    Local Article Number: NOVUNBP1-42568SS
    Specifications:
    More
    Specifications
    Antigen symbol
    TUBB3
    Antigen name
    tubulin, beta 3 class III
    Environmentally Preferable
    Antigen synonyms
    CDCBM|beta-4|CFEOM3A|FEOM3|CDCBM1|TUBB4|CFEOM3
    Antibody type
    Primary
    Clonality
    Monoclonal
    Conjugation
    Unconjugated
    Clone
    2E9
    Reactivity
    Human
    Host
    Mouse
    Gene ID
    10381
    Isotype
    IgG1
    Western blot
    Yes
    ELISA
    Yes
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice