Northern Ireland
Order Entry
Anti-HPGD Rabbit Polyclonal Antibody
Anti-HPGD Rabbit Polyclonal Antibody
 
undefined
Anti-HPGD Rabbit Polyclonal Antibody

 

  • Catalog No:
  • NOVUNB200-179SS
  • NOVUNB200-179
  • Antigen symbol:
  • 15-PGDH
  • 15-PGDH
  • Antigen name:
  • Prostaglandin dehydrogenase 1
  • Prostaglandin dehydrogenase 1
  • Antigen synonyms:
  • HPGD|PGDH1|Prostaglandin dehydrogenase 1|15-hydroxyprostaglandin dehydrogenase [NAD+]
  • HPGD|PGDH1|Prostaglandin dehydrogenase 1|15-hydroxyprostaglandin dehydrogenase [NAD+]
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • 3248
  • 3248
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

 

  •  
     
    Size
    Pk
     
     
  • NOVUNB200-179SS
    NB200-179SS
    25 µL
    25 µl
       
       
    1 * 25 µl
    1 * 25 µl - £261.00
    £261.00
    1 * 25 µl
       £261.00

     : NB200-179SS
     : NOVUNB200-179SS
     
     
    Specifications
    Antigen symbol
    15-PGDH
    Antigen name
    Prostaglandin dehydrogenase 1
    Environmentally Preferable
    Antigen synonyms
    HPGD|PGDH1|Prostaglandin dehydrogenase 1|15-hydroxyprostaglandin dehydrogenase [NAD+]
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    3248
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • NOVUNB200-179
    NB200-179
    100 µL
    0,1 mL
       
       
    1 * 0,1 mL
    1 * 0,1 mL - £611.00
    £611.00
    1 * 0,1 mL
       £611.00

     : NB200-179
     : NOVUNB200-179
     
     
    Specifications
    Antigen symbol
    15-PGDH
    Antigen name
    Prostaglandin dehydrogenase 1
    Environmentally Preferable
    Antigen synonyms
    HPGD|PGDH1|Prostaglandin dehydrogenase 1|15-hydroxyprostaglandin dehydrogenase [NAD+]
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human
    Host
    Rabbit
    Gene ID
    3248
    Isotype
    Western blot
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Storage temperature
    Cool
    Shipping temperature
    Ice