Anti-IgG Bovine Polyclonal Antibody (CF™ 633)

Supplier: Biotium

20296.EA 334 GBP
20296. 20296-1
Anti-IgG Bovine Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG (Goat) (goat IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Bovine
Isotype: none
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR