Anti-IgG2b Goat Polyclonal Antibody (CF™ 555)

Supplier: Biotium

20267-1EA 155 GBP
20267-1 20267.
Anti-IgG2b Goat Polyclonal Antibody (CF™ 555)
Antibodies
Type: Secondary
Antigen: IgG2b (Mouse) (mouse IgG2b)
Clonality: polyclonal
Clone:
Conjugation: CF® 555 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR