Anti-IgG1 Goat Polyclonal Antibody (CF™ 633)

Supplier: Biotium
20250.EA 293 GBP
20250. 20250-1
Anti-IgG1 Goat Polyclonal Antibody (CF™ 633)
Antibodies
Type: Secondary
Antigen: IgG1 (Mouse) (mouse IgG1)
Clonality: polyclonal
Clone:
Conjugation: CF® 633 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR