Northern Ireland
Order Entry
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
 :  Biotium
 
Anti-IgG Goat Polyclonal Antibody (CF™ 594)
 :  Biotium

 

  • Catalog No:
  • 20119-1
  • 20119.
  • Antigen symbol:
  • IgG
  • IgG
  • Antigen name:
  • Mouse IgG
  • Mouse IgG
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • CF® 594
  • CF® 594
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Goat
  • Goat
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Antibody fragment:
  • F(ab')2
  • F(ab')2
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • Affinity purified
  • Affinity purified
  • Concentration:
  • 2 mg/mL
  • 2 mg/mL
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
  • Storage temperature:
  • Store at –20 °C.
  • Store at –20 °C.
  • Shipping temperature:
  • Ice
  • Ice

 

 

 

  •  
     
    Size
    Pk
     
     
  • 20119-1
    20119-1
    50 uL
    50 µl
       
       
    1 * 50 µl
    1 * 50 µl - £145.00
    £145.00
    1 * 50 µl
       £145.00

     
     
    Specifications
    Antigen symbol
    IgG
    Antigen name
    Mouse IgG
    Environmentally Preferable
    Antibody type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    CF® 594
    Reactivity
    Mouse
    Host
    Goat
    Isotype
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Antibody fragment
    F(ab')2
    Form
    Liquid
    Purification
    Affinity purified
    Concentration
    2 mg/mL
    Cross adsorption
    No
    Storage buffer
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
    Storage temperature
    Store at –20 °C.
    Shipping temperature
    Ice
  • 20119.
    20119
    250 μL
    0,5 mL
       
       
    1 * 0,5 mL
    1 * 0,5 mL - £229.00
    £229.00
    1 * 0,5 mL
       £229.00

     
     
    Specifications
    Antigen symbol
    IgG
    Antigen name
    Mouse IgG
    Environmentally Preferable
    Antibody type
    Secondary
    Clonality
    Polyclonal
    Conjugation
    CF® 594
    Reactivity
    Mouse
    Host
    Goat
    Isotype
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    ImmunoFluorescence
    Yes
    Flow cytometry
    Yes
    Antibody fragment
    F(ab')2
    Form
    Liquid
    Purification
    Affinity purified
    Concentration
    2 mg/mL
    Cross adsorption
    No
    Storage buffer
    PBS, 50% glycerol, 2 mg/ml BSA, 0,05% sodium azide
    Storage temperature
    Store at –20 °C.
    Shipping temperature
    Ice