Northern Ireland
Order Entry
Anti-PASK Rabbit Polyclonal Antibody
  ABCAAB115164-100
 :  Abcam
Anti-PASK Rabbit Polyclonal Antibody
  ABCAAB115164-100
 :  Abcam
Price
£570.00
Log in for availability
Unit of Measure 1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    PAS domain containing serine/threonine kinase
  • Antigen symbol:
    PASK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG