Northern Ireland
Order Entry
Anti-ACTN3 Rabbit Polyclonal Antibody
  ABCAAB113234-50
 :  Abcam
Anti-ACTN3 Rabbit Polyclonal Antibody
  ABCAAB113234-50
 :  Abcam
Price
£409.00
Log in for availability
Unit of Measure 1 * 50 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    actinin alpha 3 (gene/pseudogene)
  • Antigen symbol:
    ACTN3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µG

 

 

Type: Primary
Antigen: ACTN3 (actinin, alpha 3 (gene/pseudogene))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human