Northern Ireland
Order Entry
Anti-IgA Goat Polyclonal Antibody
  ABCAAB112822-1
 :  Abcam
Anti-IgA Goat Polyclonal Antibody
  ABCAAB112822-1
 :  Abcam
 
£301.00
   
  1 * 1 mg

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µg
  • Pk:
    1 mg

 

 

Type: Secondary
Antigen: IgA (Dog) (dog IgA)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: