Anti-IgA Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
ABCAAB112817-1
: Abcam
- Antibody type:Secondary
- Antigen symbol:IgA
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:1 mg
Type: Secondary
Antigen: IgA (Chicken) (chicken IgA)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: