Northern Ireland
Order Entry
Anti-PYY Mouse Monoclonal Antibody [clone: 1]
Catalog # ABCAAB112474-100
Supplier:  Abcam
Anti-PYY Mouse Monoclonal Antibody [clone: 1]
  ABCAAB112474-100
 :  Abcam
 
£559.00
   
  1 * 100 µG

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Peptide YY
  • Antigen symbol:
    PYY
  • Clonality:
    Monoclonal
  • Clone:
    1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: PYY (peptide YY)
Clonality: monoclonal
Clone: 1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human