Anti-DPP10 Goat Polyclonal Antibody
Catalog # ABCAAB111985-50
Supplier: Abcam
- Antibody type:Primary
- Antigen name:Dipeptidylpeptidase 10
- Antigen symbol:DPP10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Type: Primary
Antigen: DPP10 (dipeptidyl-peptidase 10 (non-functional))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human