Northern Ireland
Order Entry
Anti-p95 NBS1 Rabbit Polyclonal Antibody
Anti-p95 NBS1 Rabbit Polyclonal Antibody
  ABCAAB111373-100
 :  Abcam
Anti-p95 NBS1 Rabbit Polyclonal Antibody
  ABCAAB111373-100
 :  Abcam
 
£591.00
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen symbol:
    P95 NBS1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: p95 NBS1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse