Type: Primary
Antigen: VCL (vinculin)
Clonality: Monoclonal
Clone: SP117
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Catalog No:
- ABCAAB111004-100
- Antigen symbol:
- VCL
- Antigen name:
- Vinculin
- Antigen synonyms:
- MV|Metavinculin|VCL
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- SP117
- Reactivity:
- Human
- Host:
- Rabbit
- Isotype:
- IgG
- ImmunoChemistry:
- Yes
- Form:
- Liquid
- Storage buffer:
- pH: 7,6
Preservative: 0,1% Sodium azide
Constituents: PBS, 1% BSA
- Storage temperature:
- Cool
- Shipping temperature:
- Ice
- Tested applications:
- ImmunoChemistry
Specifications
Product Information
- Catalog #Supplier #PkAvailabilityPrice
- ABCAAB111004-100AB111004-100100 µl1 * 100 µl£629.00
Specifications
Antigen symbolVCLAntigen nameVinculinEnvironmentally PreferableAntigen synonymsMV|Metavinculin|VCLAntibody typePrimaryClonalityMonoclonalConjugationUnconjugatedCloneSP117ReactivityHumanHostRabbitIsotypeIgGImmunoChemistryYesFormLiquidStorage bufferpH: 7,6
Preservative: 0,1% Sodium azide
Constituents: PBS, 1% BSAStorage temperatureCoolShipping temperatureIceTested applicationsImmunoChemistry