Anti-Dynamitin Rabbit Monoclonal Antibody [clone: EPR5094]
Catalog # ABCAAB109752-100
Supplier: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Dynamitin
- Clonality:Monoclonal
- Clone:EPR5094
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
Type: Primary
Antigen: Dynamitin
Clonality: monoclonal
Clone: EPR5094
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: