Anti-TBK1 Rabbit Monoclonal Antibody [clone: EPR2867]
ABCAAB109735-100
: Abcam
- Antibody type:Primary
- Antigen name:TANK-binding kinase 1
- Antigen symbol:TBK1
- Clonality:Monoclonal
- Clone:EPR2867
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: TBK1 (TANK-binding kinase 1)
Clonality: monoclonal
Clone: EPR2867
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: