Northern Ireland
Order Entry
Anti-TRIM28 Rabbit Monoclonal Antibody [clone: EPR5217]
Anti-TRIM28 Rabbit Monoclonal Antibody [clone: EPR5217]
  ABCAAB109289-100
 :  Abcam
Anti-TRIM28 Rabbit Monoclonal Antibody [clone: EPR5217]
  ABCAAB109289-100
 :  Abcam
 
£634.00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    tripartite motif containing 28
  • Antigen symbol:
    TRIM28
  • Clonality:
    Monoclonal
  • Clone:
    EPR5217
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Type: Primary
Antigen: TRIM28 (tripartite motif containing 28)
Clonality: monoclonal
Clone: EPR5217
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: