Northern Ireland
Order Entry
Anti-BCAS2 Rabbit monoclonal antibody unconjugated [clone: EPR4375]
Anti-BCAS2 Rabbit monoclonal antibody unconjugated [clone: EPR4375]
  ABCAAB108330-100
 :  Abcam
Anti-BCAS2 Rabbit monoclonal antibody unconjugated [clone: EPR4375]
  ABCAAB108330-100
 :  Abcam
 
£548.00
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    breast carcinoma amplified sequence 2
  • Antigen symbol:
    BCAS2
  • Clonality:
    Monoclonal
  • Clone:
    EPR4375
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl