Anti-CD73 Mouse Monoclonal Antibody [clone: CB-AD2] (PerCP (Peridinin-Chlorophyll Protein Complex))
ABCAAB106677-50
: Abcam
- Antibody type:Primary
- Antigen name:5'-nucleotidase, ecto
- Antigen symbol:NT5E
- Clonality:Monoclonal
- Clone:CB-AD2
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Seq:0780
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 Tests
- Pack type:Vial
- Pk:50 Tests