Northern Ireland
Order Entry
Anti-cyp19a1b Rabbit Polyclonal Antibody
Anti-cyp19a1b Rabbit Polyclonal Antibody
  ABCAAB106168-200
 :  Abcam
Anti-cyp19a1b Rabbit Polyclonal Antibody
  ABCAAB106168-200
 :  Abcam
 
£548.00
   
  1 * 200 µl

 

  • Antibody type:
    Primary
  • Antigen symbol:
    cyp19a1b
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Zebrafish
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl