Anti-UBA52 Rabbit Polyclonal Antibody
ABCAAB104455-200
: Abcam
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin A-52 Residue Ribosomal Protein Fusion product 1
- Antigen symbol:UBA52
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:200 µl