Anti-TREM1 Rabbit Polyclonal Antibody
ABCAAB104413-100
: Abcam
- Antibody type:Primary
- Antigen name:Triggering receptor expressed on myeloid cells 1
- Antigen symbol:TREM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG