Anti-IgG Rabbit Polyclonal Antibody
Catalog # ABCAAB102248-500
Supplier: Abcam
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG