Anti-ADORA2A Rabbit Polyclonal Antibody
ABCAAB101678-100
: Abcam
- Antibody type:Primary
- Antigen name:adenosine A2a receptor
- Antigen symbol:ADORA2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µG