Northern Ireland
Order Entry
Anti-IgG4 Mouse Monoclonal Antibody [clone: HP6023] (FITC (Fluorescein Isothiocyanate))
  ABCAAB99815-250
 :  Abcam
Anti-IgG4 Mouse Monoclonal Antibody [clone: HP6023] (FITC (Fluorescein Isothiocyanate))
  ABCAAB99815-250
 :  Abcam
 
£457.00
   
  1 * 250 µG

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG4
  • Clonality:
    Monoclonal
  • Clone:
    HP6023
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG3
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    250 µg
  • Pk:
    250 µG