Anti-SUGT1 Rabbit Polyclonal Antibody
ABCAAB99293-100
: Abcam
- Antibody type:Primary
- Antigen name:SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae)
- Antigen symbol:SUGT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl