Anti-IgA Goat Polyclonal Antibody (Biotin)
ABCAAB98552-500
: Abcam
- Antibody type:Secondary
- Antigen symbol:IgA
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG