Anti-IgM Goat Polyclonal Antibody (DyLight® 594)
Catalog # ABCAAB98546-500
Supplier: Abcam
Specifications
- Antibody type:Secondary
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:DyLight® 594
- Flow cytometry:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µg
- Pk:500 µG