Anti-IgM Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
ABCAAB97205-1
: Abcam
- Antibody type:Secondary
- Antigen symbol:IgM
- Clonality:Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 mg
- Pk:1 mg