Northern Ireland
Order Entry
Anti-IgA Goat Polyclonal Antibody (DyLight® 488)
  ABCAAB96995-500
 :  Abcam
Anti-IgA Goat Polyclonal Antibody (DyLight® 488)
  ABCAAB96995-500
 :  Abcam
 
£301.00
   
  1 * 500 µG

 

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgA
  • Clonality:
    Polyclonal
  • Conjugation:
    DyLight® 488
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    500 µg
  • Pk:
    500 µG