Northern Ireland
Order Entry
Anti-TBCK Rabbit Polyclonal Antibody
  ABCAAB96188-50
 :  Abcam
Anti-TBCK Rabbit Polyclonal Antibody
  ABCAAB96188-50
 :  Abcam
 
£548.00
   
  1 * 50 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    TBC1 domain containing kinase
  • Antigen symbol:
    TBCK
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    50 µl