Northern Ireland
Order Entry
Anti-Phosphoserine Aminotransferase Rabbit Polyclonal Antibody
Anti-Phosphoserine Aminotransferase Rabbit Polyclonal Antibody
  ABCAAB96136-50
 :  Abcam
undefined
Anti-Phosphoserine Aminotransferase Rabbit Polyclonal Antibody
  ABCAAB96136-50
 :  Abcam
 
£494.00
   
  1 * 50 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Phosphoserine Aminotransferase
  • Antigen symbol:
    PSAT
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Seq:
    1202
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pack type:
    Vial
  • Pk:
    50 µl