Northern Ireland
Order Entry
Anti-USP19 Rabbit Polyclonal Antibody
Anti-USP19 Rabbit Polyclonal Antibody
Catalog # ABCAAB93159-100
Supplier:  Abcam
Anti-USP19 Rabbit Polyclonal Antibody
  ABCAAB93159-100
 :  Abcam
 
£591.00
   
  1 * 100 µG

 

  • Antibody type:
    Primary
  • Antigen name:
    Ubiquitin specific peptidase 19
  • Antigen symbol:
    USP19
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

About this item