Anti-EYA2 Rabbit monoclonal antibody unconjugated [clone: EPR2942]
ABCAAB92505-100
: Abcam
- Antibody type:Primary
- Antigen name:Eyes Absent homolog 2
- Antigen symbol:EYA2
- Clonality:Monoclonal
- Clone:EPR2942
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl