Northern Ireland
Order Entry
Anti-PLAG1 Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-PLAG1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB101852-100
  • BIRBORB101852-200
  • Antigen symbol:
  • PLAG1
  • PLAG1
  • Antigen name:
  • Pleiomorphic adenoma gene 1 protein
  • Pleiomorphic adenoma gene 1 protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • liquid
  • liquid
  • Molecular weight:
  • 56 kDa
  • 56 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

 

Product Information

  • Catalog #
    Supplier #
    Size
    Pk
    Availability
    Price
  • BIRBORB101852-100
    ORB101852-100
    100 µg
    100 µG
       
    Log in for availability
    1 * 100 µG
    1 * 100 µG

     : ORB101852-100
     : BIRBORB101852-100
    Specifications:
    More
    Specifications
    Antigen symbol
    PLAG1
    Antigen name
    Pleiomorphic adenoma gene 1 protein
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    56 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB101852-200
    ORB101852-200
    200 µg
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG

     : ORB101852-200
     : BIRBORB101852-200
     
     
    Specifications
    Antigen symbol
    PLAG1
    Antigen name
    Pleiomorphic adenoma gene 1 protein
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    56 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice