Anti-PPP2R3A Rabbit Polyclonal Antibody
: Biorbyt
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunoprecipitation: 1:20-100; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: PPP2R3A (protein phosphatase 2, regulatory subunit B'', alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- BIRBORB101831-100
- BIRBORB101831-200
- Antigen symbol:
- PPP2R3A
- PPP2R3A
- Antigen name:
- protein phosphatase 2, regulatory subunit B'', alpha
- protein phosphatase 2, regulatory subunit B'', alpha
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- liquid
- liquid
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
Specifications
Antigen symbolPPP2R3AAntigen nameprotein phosphatase 2, regulatory subunit B'', alphaEnvironmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesFormliquidPurificationpurified by affinity chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIceSpecifications
Antigen symbolPPP2R3AAntigen nameprotein phosphatase 2, regulatory subunit B'', alphaEnvironmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesFormliquidPurificationpurified by affinity chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIce