Anti-KCNN4 Rabbit Polyclonal Antibody
: Biorbyt
Western Blot: 1:100-500; ELISA: 1:500-1000; Immunochemistry: 1:100-500; Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)
Type: Primary
Antigen: KCNN4 (potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- BIRBORB101130-200
- BIRBORB101130-100
- Antigen symbol:
- KCNN4
- KCNN4
- Antigen name:
- potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4
- potassium intermediate/small conductance calcium-activated channel, subfamily N, member 4
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Form:
- liquid
- liquid
- Molecular weight:
- 50 kDa
- 50 kDa
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- 0,5 mg/ml
- 0,5 mg/ml
- Storage buffer:
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Product Information
- Catalog #Supplier #SizePkAvailabilityPrice
Specifications
Antigen symbolKCNN4Antigen namepotassium intermediate/small conductance calcium-activated channel, subfamily N, member 4Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesFormliquidMolecular weight50 kDaPurificationpurified by affinity chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIceSpecifications
Antigen symbolKCNN4Antigen namepotassium intermediate/small conductance calcium-activated channel, subfamily N, member 4Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesFormliquidMolecular weight50 kDaPurificationpurified by affinity chromatographyConcentration0,5 mg/mlStorage buffer10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerolStorage temperatureCoolShipping temperatureIce