Northern Ireland
Order Entry
Anti-WNT6 Rabbit Polyclonal Antibody
Anti-WNT6 Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-WNT6 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB101095-100
  • BIRBORB101095-200
  • Antigen symbol:
  • WNT6
  • WNT6
  • Antigen name:
  • wingless-type MMTV integration site family, member 6
  • wingless-type MMTV integration site family, member 6
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • liquid
  • liquid
  • Molecular weight:
  • 40 kDa
  • 40 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

 

 

  •  
     
    Size
    Pk
     
     
  • BIRBORB101095-100
    ORB101095-100
    100 µg
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG

     
     
    Specifications
    Antigen symbol
    WNT6
    Antigen name
    wingless-type MMTV integration site family, member 6
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    40 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB101095-200
    ORB101095-200
    200 µg
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG

     
     
    Specifications
    Antigen symbol
    WNT6
    Antigen name
    wingless-type MMTV integration site family, member 6
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    40 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice