Northern Ireland
Order Entry
Anti-SART1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
Anti-SART1 Rabbit Polyclonal Antibody
 :  Biorbyt

 

  • Catalog No:
  • BIRBORB100836-100
  • BIRBORB100836-200
  • Antigen symbol:
  • SART1
  • SART1
  • Antigen name:
  • Squamous cell carcinoma antigen recognized by T cells 1
  • Squamous cell carcinoma antigen recognized by T cells 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human, Rat, Mouse
  • Human, Rat, Mouse
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • liquid
  • liquid
  • Molecular weight:
  • 90 kDa
  • 90 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Related Information

Product Information

  •  
     
    Size
    Pk
     
     
  • BIRBORB100836-100
    ORB100836-100
    100 µg
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG

     
     
    Specifications
    Antigen symbol
    SART1
    Antigen name
    Squamous cell carcinoma antigen recognized by T cells 1
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    90 kDa
    Purification
    purified by affinity chromatography
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB100836-200
    ORB100836-200
    200 µg
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG

     
     
    Specifications
    Antigen symbol
    SART1
    Antigen name
    Squamous cell carcinoma antigen recognized by T cells 1
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Human, Rat, Mouse
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    90 kDa
    Purification
    purified by affinity chromatography
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice