Northern Ireland
Order Entry
Anti-VEGFA Rabbit Polyclonal Antibody
Anti-VEGFA Rabbit Polyclonal Antibody
 :  Biorbyt
 
undefined
Anti-VEGFA Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB100468-100
  • BIRBORB100468-200
  • Antigen symbol:
  • VEGFA
  • VEGFA
  • Antigen name:
  • Vascular endothelial growth factor A
  • Vascular endothelial growth factor A
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Form:
  • liquid
  • liquid
  • Molecular weight:
  • 40 - 45 kDa
  • 40 - 45 kDa
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Storage buffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

 

  •  
     
    Size
    Pk
     
     
  • BIRBORB100468-100
    ORB100468-100
    100 µg
    100 µG
       
       
    1 * 100 µG
    1 * 100 µG

     
     
    Specifications
    Antigen symbol
    VEGFA
    Antigen name
    Vascular endothelial growth factor A
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    40 - 45 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice
  • BIRBORB100468-200
    ORB100468-200
    200 µg
    200 µG
       
       
    1 * 200 µG
    1 * 200 µG

     
     
    Specifications
    Antigen symbol
    VEGFA
    Antigen name
    Vascular endothelial growth factor A
    Environmentally Preferable
    Antibody type
    Primary
    Clonality
    Polyclonal
    Conjugation
    Unconjugated
    Reactivity
    Host
    Rabbit
    Isotype
    IgG
    Western blot
    Yes
    ELISA
    Yes
    ImmunoChemistry
    Yes
    Form
    liquid
    Molecular weight
    40 - 45 kDa
    Purification
    purified by affinity chromatography
    Concentration
    0,5 mg/ml
    Storage buffer
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
    Storage temperature
    Cool
    Shipping temperature
    Ice