Anti-ELL2 Rabbit polyclonal antibody
BIRBORB66958-100
: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:elongation factor, RNA polymerase II, 2
- Antigen symbol:ELL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Western blot:Yes
- Storage buffer:1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:C-terminal region of human ELL2 [distinct from the antigen for R0211-1, ELL2 (C-term1) antibody]
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
Western Blot: 1:1000-1:5000
Type: Primary
Antigen: ELL2 (elongation factor, RNA polymerase II, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: