Northern Ireland
Order Entry
Anti-AURKB Rabbit Polyclonal Antibody
Catalog # BIRBORB66849-100
Supplier:  Biorbyt
undefined
Anti-AURKB Rabbit Polyclonal Antibody
Catalog # BIRBORB66849-100
Supplier:  Biorbyt
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    aurora kinase B
  • Antigen symbol:
    AURKB
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage buffer:
    1 x PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    N-terminal region of human AURKB
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

About this item

Western Blot: 1:1000-1:5000

Type: Primary
Antigen: AURKB (aurora kinase B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: