Northern Ireland
Order Entry
Anti-HDAC2 Rabbit polyclonal antibody
Anti-HDAC2 Rabbit polyclonal antibody
  BIRBORB66724-100
 :  Biorbyt
undefined
Anti-HDAC2 Rabbit polyclonal antibody
  BIRBORB66724-100
 :  Biorbyt
 
 
   
  1 * 100 µl

 

  • Antibody type:
    Primary
  • Antigen name:
    Histone deacetylase 2
  • Antigen symbol:
    HDAC2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage buffer:
    PBS (pH 7,4), 40% Glycerol, 0,02% Thimerosal.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Western Blot: 1:1000-1:5000

Type: Primary
Antigen: HDAC2 (histone deacetylase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: