Anti-ATP6AP1 Rabbit polyclonal antibody
: Biorbyt
Type: Primary
Antigen: ATP6AP1 (ATPase, H+ transporting, lysosomal accessory protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
- Catalog No:
- BIRBORB49112-200
- BIRBORB49112-100
- Antigen symbol:
- ATP6AP1
- ATP6AP1
- Antigen name:
- ATPase, H+ transporting, lysosomal accessory Protein 1
- ATPase, H+ transporting, lysosomal accessory Protein 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- SizePk
Specifications
Antigen symbolATP6AP1Antigen nameATPase, H+ transporting, lysosomal accessory Protein 1Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesPurificationpurified by affinity chromatographyStorage temperatureCoolShipping temperatureIceSpecifications
Antigen symbolATP6AP1Antigen nameATPase, H+ transporting, lysosomal accessory Protein 1Environmentally PreferableAntibody typePrimaryClonalityPolyclonalConjugationUnconjugatedReactivityHuman, Rat, MouseHostRabbitIsotypeIgGWestern blotYesImmunoChemistryYesPurificationpurified by affinity chromatographyStorage temperatureCoolShipping temperatureIce