Anti-Pro-BNP Mouse Monoclonal Antibody
BIRBORB48855-200
: Biorbyt
- Antibody type:Primary
- Antigen symbol:Pro-BNP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Peptide
- Size:200 µg
- Pk:200 µG
Type: Primary
Antigen: Pro-BNP
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity: Human