Northern Ireland
Order Entry
Anti-POMC Rabbit Polyclonal Antibody
  BIRBORB48725-200
 :  Biorbyt
Anti-POMC Rabbit Polyclonal Antibody
Catalog # BIRBORB48725-200
Supplier:  Biorbyt
 
 
   
  1 * 200 µG

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    proopiomelanocortin
  • Antigen symbol:
    POMC
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Format:
    ascites fluid
  • Storage buffer:
    0,03% sodium azide.
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Peptide
  • Purification:
    purified by affinity chromatography
  • Size:
    200 µg
  • Pk:
    200 µG

Specifications

About this item

Type: Primary
Antigen: POMC (proopiomelanocortin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat